Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-15 Antibody (OTI2B4), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP271034C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271034C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271034C Supplier Novus Biologicals Supplier No. NBP271034C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-15 Monoclonal specifically detects IL-15 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-15
Applications Western Blot
Classification Monoclonal
Clone OTI2B4
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias IL-15MGC9721, interleukin 15, interleukin-15
Gene Symbols IL15
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 49-162 of human IL-15 (NP_751914) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3600.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.