Learn More
Invitrogen™ IL-15 Monoclonal Antibody (ct2nu), Functional Grade, eBioscience™, Invitrogen™

Description
Description: The ct2nu monoclonal antibody reacts with human IL-15 and neutralizes its bioactivity. IL-15, or IL-T, is a 14 kDa proinflammatory cytokine and member of the IL-2 family. It plays a role in the development and maturation of macrophages and dendritic cells, and is essential to the proliferation and survival of NK cells and CD8^+ T cells. IL-15 is expressed by antigen-presenting cells, epithelial cells, and stromal cells. IL-15 activates the Jak/STAT pathway through its heterotrimeric receptor. Two subunits of this receptor, the beta and gamma chains, are shared with the rest of the family, while the alpha subunit is unique to IL-15. In mice, the alpha chain is required to chaperone IL-15 from the ER to secretion. This chaperoning is not necessary in humans, and free IL-15 is detectable in human biological samples. However, the receptor-cytokine complex has been indentified in humans and demonstrates agonistic activity several-fold higher than IL-15 alone. Applications Reported: The monoclonal antibody ct2nu reacts with and inhibits the bioactivity of human IL-15. Applications Tested: The ND50 of this antibody, as measured by inhibition of proliferation in CTLL-2 cells, is 1-10 ng/mL in the presence of 0.5 ng/mL recombit human IL-15. Neutralization dose will vary based on cell type, cytokine concentration, and assay method. This antibody should be carefully titrated for optimal performance in the assay of interest. Endotoxin: Less than 0.
Specifications
Specifications
| Antigen | IL-15 |
| Applications | Functional Assay, Neutralization |
| Classification | Monoclonal |
| Clone | ct2nu |
| Concentration | 1 mg/mL |
| Conjugate | Functional Grade |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | IL15 |
| Gene Accession No. | P40933 |
| Gene Alias | AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15 |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.