Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-15R alpha Antibody (2639B), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB339884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339884 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339884 Supplier Novus Biologicals Supplier No. FAB10900MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-15R alpha Monoclonal antibody specifically detects IL-15R alpha in Human samples. It is validated for Flow Cytometry

Specifications

Antigen IL-15R alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 2639B
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human IL-15R alpha, Met1-Lys173, Accession # NP_751951
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3601
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.