Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-15R alpha Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB200812 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200812 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200812 Supplier Novus Biologicals Supplier No. NBP241212F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-15R alpha Polyclonal antibody specifically detects IL-15R alpha in Human, Mouse, Rat samples. It is validated for ELISA

Specifications

Antigen IL-15R alpha
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the center of human IL-15RA. The immunogen is located within amino acids 40 - 90 of IL-15RA. Amino Acid Squence: NSGFKRKAGTSSLTE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3601
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.