Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-15R alpha Antibody (RM0103-6C38), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB206070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB206070 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB206070 Supplier Novus Biologicals Supplier No. NBP122466JF646
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-15R alpha Monoclonal antibody specifically detects IL-15R alpha in Mouse samples. It is validated for Western Blot

Specifications

Antigen IL-15R alpha
Applications Western Blot
Classification Monoclonal
Clone RM0103-6C38
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179
Host Species Rat
Immunogen Mouse recombinant IL-15R alpha (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3601
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.