Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-16 Antibody (70720), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB304112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304112 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304112 Supplier Novus Biologicals Supplier No. FAB3161A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-16 Monoclonal specifically detects IL-16 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen IL-16
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 70720
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias FLJ16806, FLJ42735, FLJ44234, HsT19289, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-16 isoform 1, Met1203-Ser1332, Accession # Q14005
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cytokine Research, Hypoxia, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3603
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.