Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17/IL-17A Antibody (41809) [DyLight 680], Novus Biologicals™
SDP

Catalog No. NB372837 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372837 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372837 Supplier Novus Biologicals Supplier No. FAB317Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human, Primate samples. It is validated for ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Immunohistochemistry, Immunoprecipitation, Neutralization.

Specifications

Antigen IL-17/IL-17A
Applications ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Immunohistochemistry, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 41809
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunoprecipitation, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Formulation 50mM Sodium Borate
Gene Alias CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-17, Ile20-Ala155, Accession # Q16552
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3605
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.