Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17/IL-17A Antibody (4k5F6), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP224765UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224765UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224765UV Supplier Novus Biologicals Supplier No. NBP224765UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-17/IL-17A
Applications Western Blot
Classification Monoclonal
Clone 4k5F6
Conjugate DyLight 350
Dilution Western Blot
Gene Accession No. Q6NZ94
Gene Alias CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Gene Symbols IL17A
Host Species Mouse
Immunogen A synthetic peptide between amino acids 1-75 of human IL-17A was used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3605
Test Specificity The amino acid sequence used as immunogen is 100% homologous in Human, and 94% in Mouse, Rat, Monkey, Opossum, Horse, Cow and Chimpanzee S1PR1.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.