Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17/IL-17A Antibody (4k5F6), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB337302 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337302 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337302 Supplier Novus Biologicals Supplier No. NBP224765MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Human samples. It is validated for Western Blot

Specifications

Antigen IL-17/IL-17A
Applications Western Blot
Classification Monoclonal
Clone 4k5F6
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Host Species Mouse
Immunogen A synthetic peptide between amino acids 1-75 of human IL-17A was used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3605
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.