Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17/IL-17A Antibody (TC11-18H10), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP172027R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP172027R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP172027R Supplier Novus Biologicals Supplier No. NBP172027R
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IL-17/IL-17A
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TC11-18H10
Concentration 0.5 mg/ml
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Gene Symbols IL17A
Host Species Rat
Immunogen E. coli-expressed, recombinant mouse IL-17/IL-17A
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3605.0
Test Specificity Recognizes mouse Interleukin 17, a 32kDa disulfide-linked homodimer consisting of two 136 amino acid chains, which belong to the IL-17 family (classified as IL-17A to IL-17F), and signal through the IL-17 receptor (IL-17R/CDw217).
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.