Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17/IL-17A Mouse anti-Non-human Primate, Clone: MT241, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318299250UG
This item is not returnable.
View return policy
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Non-human Primate samples. It is validated for ELISA, ELISPOTSpecifications
IL-17/IL-17A | |
Monoclonal | |
0.5 mg/ml | |
0.2 um filtered PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Non-human Primate | |
Purified |
ELISA | |
MT241 | |
Unconjugated | |
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
Recombinant human IL-17/ IL-17A | |
250 μg | |
Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
3605 | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction