Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17/IL-17A Mouse anti-Non-human Primate, Clone: MT241, Novus Biologicals™

Mouse Monoclonal Antibody
$286.00 - $1033.00
Specifications
| Antigen | IL-17/IL-17A |
|---|---|
| Clone | MT241 |
| Concentration | 0.5 mg/ml |
| Applications | ELISA |
| Classification | Monoclonal |
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Non-human Primate samples. It is validated for ELISA, ELISPOTSpecifications
| IL-17/IL-17A | |
| 0.5 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Non-human Primate | |
| CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| Recombinant human IL-17/ IL-17A | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| MT241 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
| 0.2 um filtered PBS | |
| 3605 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title