Learn More
IL-17AF Monoclonal Antibody (1402nAF), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$3260.00
Specifications
Antigen | IL-17AF |
---|---|
Clone | 1402nAF |
Concentration | 1.0 mg/mL |
Applications | Functional Assay, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-917-238
![]() |
Invitrogen™
16917238 |
5 mg |
Each of 1 for $3,260.00
|
|
|||||
Description
Description: The 1402nAF monoclonal antibody reacts with mouse interleukin (IL)-17AF heterodimer. This antibody specifically reacts with this heterodimer and has no reactivity for IL-17A or IL-17F homodimers. Applications Reported: This 1402nAF antibody has been reported for use as a neutralization antibody of IL-17AF biological activity. Applications Tested: The Functional Grade purified format of the 1402nAF antibody has been tested for neutralization of IL-17AF biological activity. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
IL-17A and IL-17F are well-characterized homodimeric cytokines secreted by T helper 17 (Th17) cells, gamma-delta T cells and several subsets of innate lymphoid cells. Somewhat less appreciated, IL-17A and IL-17F subunits can also form the heterodimer, IL-17AF. Together, these three dimers signal through the IL-17RA/IL-17RC receptor complex to mediate immune responses at mucosal interfaces and are found at lesion sites in inflammatory bowel disease, asthma, atopic dermatitis and rheumatoid arthritis.Specifications
IL-17AF | |
1.0 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
16171, 257630 | |
Primary | |
4°C, do not freeze |
1402nAF | |
Functional Assay, Neutralization | |
Functional Grade | |
Rat | |
Mouse | |
Q62386, Q7TNI7 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.