Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17B Antibody (B-B58), PE, Novus Biologicals™
SDP

Catalog No. NB340587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340587 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340587 Supplier Novus Biologicals Supplier No. NBP318115PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17B Monoclonal antibody specifically detects IL-17B in Human samples. It is validated for ELISA

Specifications

Antigen IL-17B
Applications ELISA
Classification Monoclonal
Clone B-B58
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20
Host Species Mouse
Immunogen Recombinant human IL-17B. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 27190
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.