Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17C Antibody, Novus Biologicals™
SDP

Catalog No. NBP227100SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227100SS 0.025 mg
NBP227100 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227100SS Supplier Novus Biologicals Supplier No. NBP227100SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-17C Polyclonal specifically detects IL-17C in Human, Primate samples. It is validated for Western Blot.

Specifications

Antigen IL-17C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9P0M4
Gene Alias CX2, Cytokine CX2, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401
Gene Symbols IL17C
Host Species Rabbit
Immunogen A portion of amino acids 80-130 from human IL-17C was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 27189
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, common chimpanzee, sumatran orangutan, rhesus monkey and white-cheeked gibbon, 94% in cattle, new world monkey, dog, and horse, 89% in guinea pig, 83% in wild turkey, mouse, and rat, 78% in zebra finch, and Chinese hamster, 72% in opossum, and western clawed frog, and 66% in platypus.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.