Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17C Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen IL-17C
Concentration 0.5 mg/mL
Dilution Western Blot 3-5ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227100SS
SDP
View Documents
Novus Biologicals
NBP227100SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227100
SDP
View Documents
Novus Biologicals
NBP227100
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

IL-17C Polyclonal specifically detects IL-17C in Human, Primate samples. It is validated for Western Blot.
Specifications

Specifications

IL-17C
Western Blot 3-5ug/ml
Polyclonal
Purified
RUO
Human, Primate
Q9P0M4
27189
A portion of amino acids 80-130 from human IL-17C was used as the immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Cytokine Research
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
CX2, Cytokine CX2, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401
IL17C
IgG
Protein A purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, common chimpanzee, sumatran orangutan, rhesus monkey and white-cheeked gibbon, 94% in cattle, new world monkey, dog, and horse, 89% in guinea pig, 83% in wild turkey, mouse, and rat, 78% in zebra finch, and Chinese hamster, 72% in opossum, and western clawed frog, and 66% in platypus.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.