Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17D Antibody (246018) [PE], Novus Biologicals™
SDP

Catalog No. NB399840 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399840 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399840 Supplier Novus Biologicals Supplier No. FAB15041P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17D Monoclonal specifically detects IL-17D in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot.

Specifications

Antigen IL-17D
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 246018
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias FLJ30846, IL-17Dinterleukin 27, IL-22, IL-27interleukin-17D, IL27interleukin-27, interleukin 17D, Interleukin-27
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-17D, Ala18-Pro202, Accession # Q8TAD2.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 53342
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.