Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17E/IL-25 Antibody (182203) [DyLight 350], Novus Biologicals™
SDP

Catalog No. NB400505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400505 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400505 Supplier Novus Biologicals Supplier No. FAB1258D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Human samples. It is validated for Western Blot, ELISA, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen IL-17E/IL-25
Applications Western Blot, ELISA, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 182203
Conjugate DyLight 350
Dilution Western Blot, ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias IL17E, IL-17E, IL-25, interleukin 25, Interleukin-17E, interleukin-25
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-17E, Tyr33-Gly177, Accession # Q9H293
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 64806
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.