Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17E/IL-25 Antibody (207702), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB304987 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304987 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304987 Supplier Novus Biologicals Supplier No. FAB13991S

Rat Monoclonal Antibody

IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Mouse samples. It is validated for ELISA Standard (Matched Pair), ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow Cytometry, CyTOF

Specifications

Antigen IL-17E/IL-25
Applications ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry, CyTOF
Classification Monoclonal
Clone 207702
Conjugate Alexa Fluor 750
Dilution ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias IL17E, IL-17E, IL-25, interleukin 25, Interleukin-17E, interleukin-25
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-17E/IL-25, Val17-Ala169, Accession # NP_542767
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 64806
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.