Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17E/IL-25 Antibody (68C1039.2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10056541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056541 0.1 mg
NB10056541T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056541 Supplier Novus Biologicals Supplier No. NB10056541
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 4 publications

IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human, Mouse samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IL-17E/IL-25
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 68C1039.2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:250, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 35359953), Immunohistochemistry-Paraffin reported in scientific literature (Terrier B et al)
Gene Accession No. Q9H293
Gene Alias IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25
Gene Symbols IL25
Host Species Mouse
Immunogen Amino acids 115-132 of human IL-17E (isoform CRA_a) were used as immunogen for this antibody.
Molecular Weight of Antigen 20 kDa
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64806
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.