Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17E/IL-25 Antibody (68C1039.2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
$206.50 - $499.50
Specifications
Antigen | IL-17E/IL-25 |
---|---|
Clone | 68C1039.2 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100 - 1:250, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 35359953), Immunohistochemistry-Paraffin reported in scientific literature (Terrier B et al) |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056541T
![]() |
Novus Biologicals
NB10056541SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056541
![]() |
Novus Biologicals
NB10056541 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-17E/IL-25 | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Mouse | |
IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25 | |
IL25 | |
IgG1 | |
Protein G purified | |
20 kDa |
68C1039.2 | |
Western Blot 1:100 - 1:250, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 35359953), Immunohistochemistry-Paraffin reported in scientific literature (Terrier B et al) | |
Monoclonal | |
Purified | |
RUO | |
Q9H293 | |
64806 | |
Amino acids 115-132 of human IL-17E (isoform CRA_a) were used as immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title