Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17E/IL-25 Antibody (8B1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP227324IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227324IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227324IR Supplier Novus Biologicals Supplier No. NBP227324IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen IL-17E/IL-25
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 8B1
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry
Gene Accession No. Q9H293
Gene Alias IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25
Gene Symbols IL25
Host Species Mouse
Immunogen Full-length human IL17E recombinant protein was used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 64806
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.