Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17E/IL-25 Mouse anti-Human, Clone: IL25/625, Novus Biologicals™
SDP

Catalog No. NB177503 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177503 20 μg
NB177502 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177503 Supplier Novus Biologicals Supplier No. NBP31377320UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17E/IL-25 Monoclonal antibody specifically detects IL-17E/IL-25 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen IL-17E/IL-25
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IL25/625
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias IL17E, IL-17E, IL-25, interleukin 25, Interleukin-17E, interleukin-25
Host Species Mouse
Immunogen Recombinant fragment of human IL-17E/IL-25 protein (exact sequence is proprietary) (Uniprot: IDQ9H293)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 64806
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.