Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17F Antibody (1058A), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB313821 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313821 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313821 Supplier Novus Biologicals Supplier No. FAB20571PECY7
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-17F Monoclonal specifically detects IL-17F in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-17F
Applications Flow Cytometry
Classification Monoclonal
Clone 1058A
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F
Host Species Rabbit
Immunogen E.coli-derived recombinant mouse IL-17F, Ala21-Val153, Accession # Q7TNI7
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 112744
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.