Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17F Antibody (4H450), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP227319UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227319UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227319UV Supplier Novus Biologicals Supplier No. NBP227319UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-17F
Applications Western Blot
Classification Monoclonal
Clone 4H450
Conjugate DyLight 350
Dilution Western Blot
Gene Accession No. Q96PD4
Gene Alias Cytokine ML-1, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24
Gene Symbols IL17F
Host Species Mouse
Immunogen Full-length recombinant MRkIPkVGHT FFQkPESCPP VPGGSMkLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGkEDISMN SVPIQQETLV VRRkHQGCSV SFQLEkVLVT VGCTCVTPVI HHVQ
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 112744
Test Specificity This antibody is reactive to Human.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.