Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17F Mouse anti-Human, Clone: B-F60, Novus Biologicals™
SDP

Catalog No. NB158377 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB158377 200 μg
NB158378 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB158377 Supplier Novus Biologicals Supplier No. NBP314589200UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17F Monoclonal antibody specifically detects IL-17F in Human samples. It is validated for ELISA, Functional Assay, ELISPOT

Specifications

Antigen IL-17F
Applications ELISA, Functional Assay
Classification Monoclonal
Clone B-F60
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Functional, ELISPOT
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F
Host Species Mouse
Immunogen Recombinant human IL-17F
Purification Method Affinity purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 112744
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.