Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17F Rabbit anti-Mouse, Clone: 8F5.1A9, Novus Biologicals™
SDP

Catalog No. NB153538 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153538 0.025 mg
NB153539 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB153538 Supplier Novus Biologicals Supplier No. NBP3120340.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-17F Monoclonal antibody specifically detects IL-17F in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen IL-17F
Applications Flow Cytometry
Classification Monoclonal
Clone 8F5.1A9
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry
Formulation PBS
Gene Alias Cytokine ML-1, IL-17Finterleukin-17F, interleukin 17F
Host Species Rabbit
Immunogen This antibody was raised by immunising mice with murine IL-17F coupled to ovalbumin to generate a highly immunogenic complex.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 112744
Target Species Mouse
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.