Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RA/IL-17R Antibody (49M4D2), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP225271FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225271FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225271FR Supplier Novus Biologicals Supplier No. NBP225271FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17RA/IL-17R Monoclonal specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IL-17RA/IL-17R
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 49M4D2
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Accession No. NP_055154
Gene Alias CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
Gene Symbols IL17RA
Host Species Mouse
Immunogen Within the range of amino acids 186-267 of human IL-17RA were used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23765
Test Specificity This product reacts with Human.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.