Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RA/IL-17R Antibody (657603), PerCP, Novus Biologicals™
SDP

Catalog No. NB370020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370020 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370020 Supplier Novus Biologicals Supplier No. FAB4481C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-17RA/IL-17R Monoclonal specifically detects IL-17RA/IL-17R in Mouse samples. It is validated for CyTOF, Flow Cytometry, Neutralization.

Specifications

Antigen IL-17RA/IL-17R
Applications CyTOF, Flow Cytometry, Neutralization
Classification Monoclonal
Clone 657603
Conjugate PerCP
Dilution Flow Cytometry, Neutralization, CyTOF-ready
Formulation PBS
Gene Alias CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RA/IL-17 R, Ser32-Trp322, Accession # Q60943
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 23765
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.