Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RA/IL-17R Rabbit anti-Mouse, Rat, Clone: 6Q2U9, Novus Biologicals™

Rabbit Monoclonal Antibody
$196.25 - $486.55
Specifications
| Antigen | IL-17RA/IL-17R |
|---|---|
| Clone | 6Q2U9 |
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Mouse, Rat samples. It is validated for Western BlotSpecifications
| IL-17RA/IL-17R | |
| Western Blot 1:500 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
| A synthetic peptide corresponding to a sequence within amino acids 767-866 of human IL-17RA/IL-17R (Q96F46). GCSRPAMVLTDPHTPYEEEQRQSVQSDQGYISRSSPQPPEGLTEMEEEEEEEQDPGKPALPLSPEDLESLRSLQRQLLFRQLQKNSGWDTMGSESEGPSA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 6Q2U9 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Innate Immunity | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 23765 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title