Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RA/IL-17R Rabbit anti-Mouse, Rat, Clone: 6Q2U9, Novus Biologicals™
SDP

Catalog No. NB168246 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168246 20 μg
NB168245 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168246 Supplier Novus Biologicals Supplier No. NBP31657720UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen IL-17RA/IL-17R
Applications Western Blot
Classification Monoclonal
Clone 6Q2U9
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 767-866 of human IL-17RA/IL-17R (Q96F46). GCSRPAMVLTDPHTPYEEEQRQSVQSDQGYISRSSPQPPEGLTEMEEEEEEEQDPGKPALPLSPEDLESLRSLQRQLLFRQLQKNSGWDTMGSESEGPSA
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 23765
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.