Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RB Antibody (938951), Alexa Fluor 405, Novus Biologicals™
SDP

Catalog No. MAB12074AFB Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB12074AFB 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB12074AFB Supplier Novus Biologicals Supplier No. MAB12074AF405

Mouse Monoclonal Antibody

IL-17RB Monoclonal specifically detects IL-17RB in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-17RB
Applications Flow Cytometry
Classification Monoclonal
Clone 938951
Conjugate Alexa Fluor 405
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, EVI27IL-17B receptor, IL-17 receptor B, IL17BRinterleukin 17B receptor, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor
Gene Symbols IL17RB
Host Species Mouse
Immunogen Partial recombinant human IL-17RB protein (aa18-289)
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 55540
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.