Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RB Antibody (MM0374-5P14), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB344396 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344396 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344396 Supplier Novus Biologicals Supplier No. NBP211673MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17RB Monoclonal antibody specifically detects IL-17RB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization

Specifications

Antigen IL-17RB
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Classification Monoclonal
Clone MM0374-5P14
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, EVI27IL-17B receptor, IL-17 receptor B, IL17BRinterleukin 17B receptor, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor
Host Species Mouse
Immunogen Human recombinant IL-17RB (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 55540
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.