Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RC Antibody (309822), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB313632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313632 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313632 Supplier Novus Biologicals Supplier No. FAB22691APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17RC Monoclonal specifically detects IL-17RC in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-17RC
Applications Flow Cytometry
Classification Monoclonal
Clone 309822
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias FLJ95963, FLJ96005, IL-17 receptor C, IL17F receptor, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-17 RC, Leu21-Ala454, Accession # NP_116121
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84818
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.