Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RD/SEF Antibody (309511), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB365794 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365794 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365794 Supplier Novus Biologicals Supplier No. FAB22751L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17RD/SEF Monoclonal specifically detects IL-17RD/SEF in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen IL-17RD/SEF
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 309511
Conjugate DyLight 550
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ35755, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, Sef homolog, SEFhSef
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-17RD, Ala27-Arg299, Accession # Q8NFM7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54756
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.