Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RE Antibody (934832), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB310858 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310858 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310858 Supplier Novus Biologicals Supplier No. FAB8358PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-17RE Monoclonal antibody specifically detects IL-17RE in Human samples. It is validated for Flow Cytometry

Specifications

Antigen IL-17RE
Applications Flow Cytometry
Classification Monoclonal
Clone 934832
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias IL-17 receptor E, interleukin 17 receptor E
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human IL-17 RE, Met1-His454, Accession # Q8NFR9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132014
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.