Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RE Antibody (944904), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB373767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373767 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373767 Supplier Novus Biologicals Supplier No. FAB7997W
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-17RE Monoclonal antibody specifically detects IL-17RE in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen IL-17RE
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 944904
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias IL-17 receptor E, interleukin 17 receptor E
Host Species Rat
Immunogen Mouse myeloma cell line NSO-derived recombinant mouse IL-17 RE, Met1-His414, Accession # Q8BH06
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132014
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.