Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-18 BPa/IL18BP Antibody (MM0379-10G36), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB211678647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211678647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211678647 Supplier Novus Biologicals Supplier No. NBP211678AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-18 BPa/IL18BP Monoclonal specifically detects IL-18 BPa/IL18BP in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-18 BPa/IL18BP
Applications Western Blot
Classification Monoclonal
Clone MM0379-10G36
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias IL-18BP, IL18BPa, interleukin 18 binding protein, interleukin-18-binding protein, MC51L-53L-54L homolog gene product, tadekinig-alfa
Gene Symbols IL18BP
Host Species Mouse
Immunogen Human recombinant IL-18 BPa/IL18BP
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 10068
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.