Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-18/IL-1F4 Antibody (167), Biotin, Novus Biologicals™
SDP

Catalog No. NB073000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB073000 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB073000 Supplier Novus Biologicals Supplier No. NBP290394B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen IL-18/IL-1F4
Applications ELISA
Classification Monoclonal
Clone 167
Conjugate Biotin
Dilution ELISA, Sandwich ELISA Detection
Formulation PBS
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-18/IL-1F4 (P70380; Asn36-Ser192).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3606
Test Specificity No cross-reactivity in ELISA with: Human IL1F7 Human IL1F9
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.