Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-18/IL-1F4 Antibody (925008), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB364797 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364797 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364797 Supplier Novus Biologicals Supplier No. FAB2548Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-18/IL-1F4 Monoclonal antibody specifically detects IL-18/IL-1F4 in Human samples. It is validated for ELISA, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen IL-18/IL-1F4
Applications ELISA, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 925008
Conjugate DyLight 680
Dilution ELISA, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Host Species Mouse
Immunogen E. coli-derived recombinant Rhesus monkey IL-18/IL-1F4, Tyr37-Asp193, Accession # Q14116
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Endocrinology, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3606
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.