Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-18/IL-1F4 Rabbit anti-Human, Clone: 11, Novus Biologicals™
SDP

Catalog No. NB020039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020039 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB020039 Supplier Novus Biologicals Supplier No. NBP28927750UL

Rabbit Monoclonal Antibody

IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for ELISA, Sandwich ELISA Capture.

Specifications

Antigen IL-18/IL-1F4
Applications ELISA
Classification Monoclonal
Clone 11
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Host Species Rabbit
Immunogen Recombinant Human IL-18/IL-1F4 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3606
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.