Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-18/IL-1F4, Rabbit anti-Human, Clone: 125-2H, Novus Biologicals™
SDP

Catalog No. NB014622 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014622 0.025 mg
NB014623 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB014622 Supplier Novus Biologicals Supplier No. NBP2810400.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Neutralization.

Specifications

Antigen IL-18/IL-1F4
Applications ELISA, Immunofluorescence, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 125-2H
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Neutralization
Formulation PBS
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Gene Symbols IL18
Host Species Rabbit
Immunogen Fusion of splenocytes from mice immunized with human IL-18/IL-1F4, with SP2/O mouse myeloma cells.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3606
Test Specificity Recognizes the active form of human IL-18/IL-1F4.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.