Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-18 R beta/IL-1 R7/ACPL Antibody (122), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB066214 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB066214 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB066214 Supplier Novus Biologicals Supplier No. NBP289316AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-18 R beta/IL-1 R7/ACPL Monoclonal specifically detects IL-18 R beta/IL-1 R7/ACPL in Human samples. It is validated for Sandwich ELISA Detection.

Specifications

Antigen IL-18 R beta/IL-1 R7/ACPL
Applications ELISA
Classification Monoclonal
Clone 122
Conjugate Alexa Fluor 488
Dilution Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias Accessory protein-like, ACPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-18 receptor accessory protein, IL-18 receptor beta, IL-18RAcP, IL18RB, IL-18Rbeta, IL-18R-beta, IL-1R accessory protein-like, IL1R7, IL-1R7, IL-1R-7, IL-1RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-18 R beta/IL-1 R7/ACPL (NP_003844.1; Met1-Gly357).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8807
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.