Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-18R alpha/IL-1 R5 Antibody (112614), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB174983 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174983 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174983 Supplier Novus Biologicals Supplier No. FAB1216D
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-18R alpha/IL-1 R5 Monoclonal antibody specifically detects IL-18R alpha/IL-1 R5 in Mouse samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen IL-18R alpha/IL-1 R5
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 112614
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha/IL-1 R5, Ser19-Gly326, Accession # Q61098
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8809
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.