Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-18R alpha/IL-1 R5 Antibody (70625), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB346096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346096 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346096 Supplier Novus Biologicals Supplier No. FAB840MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-18R alpha/IL-1 R5 Monoclonal specifically detects IL-18R alpha/IL-1 R5 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Neutralization.

Specifications

Antigen IL-18R alpha/IL-1 R5
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 70625
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD218a;CDw218a;IL18R1;IL18RA;IL-18Ralpha;IL-18R-alpha;IL18Ralpha2;IL1RRP;IL-1Rrp;Interleukin-18 receptor 1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-18 R alpha/IL-1 R5, Glu20-Arg329, Accession # Q13478
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8809
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.