Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-19 Antibody (152112), HRP, Novus Biologicals™
SDP

Catalog No. NB305713 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305713 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305713 Supplier Novus Biologicals Supplier No. FAB1035H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-19 Monoclonal antibody specifically detects IL-19 in Human samples. It is validated for Neutralization, Flow Cytometry, CyTOF

Specifications

Antigen IL-19
Applications Neutralization, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 152112
Conjugate HRP
Dilution Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias IL-10C, IL-19, interleukin 19, MDA1, melanoma differentiation associated protein-like protein, NG.1Melanoma differentiation-associated protein-like protein, ZMDA1interleukin-19
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-19, Leu25-Ala177, Accession # AAN40906
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 29949
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.