Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1ra/IL-1F3/IL1RN Antibody (005), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB087633 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087633 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087633 Supplier Novus Biologicals Supplier No. NBP289195IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IL-1ra/IL-1F3/IL1RN
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the human IL-1ra/IL-1F3/IL1RN.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3557
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.