Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1ra/IL-1F3/IL1RN Antibody (OTI2B1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP271022A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271022A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271022A32 Supplier Novus Biologicals Supplier No. NBP271022AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen IL-1ra/IL-1F3/IL1RN
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2B1
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4
Gene Symbols IL1RN
Host Species Mouse
Immunogen Full length recombinant protein of human IL-1ra/IL-1F3/IL1RN (NP_776215) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3557
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.