Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1ra/IL-1F3/IL1RN Antibody (OTI2B1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289192 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289192 Supplier Novus Biologicals Supplier No. NBP271022APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-1ra/IL-1F3/IL1RN Monoclonal antibody specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen IL-1ra/IL-1F3/IL1RN
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2B1
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4
Host Species Mouse
Immunogen Full length human recombinant protein of human IL1RN (NP_776215) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3557
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.