Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-2 Antibody (5334R), Biotin, Novus Biologicals™
SDP

Catalog No. NB304650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304650 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304650 Supplier Novus Biologicals Supplier No. FAB202RB
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-2 Monoclonal specifically detects IL-2 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, Neutralization.

Specifications

Antigen IL-2
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry, Neutralization
Classification Monoclonal
Clone 5334R
Conjugate Biotin
Dilution Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation PBS
Gene Alias aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-2, Ala21-Thr153, Accession # NP_000577
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3558
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.